(±)-1-Bromo-2-methylbutane ≥97% stabilised

Supplier: Thermo Fisher Scientific
Danger


Stabilization: Stabilised with potassium carbonate

H27078.14 H27078.06
ALFAH27078.14EA 0 SGD
ALFAH27078.14 ALFAH27078.06
(±)-1-Bromo-2-methylbutane ≥97% stabilised
(±)-1-Bromo-2-methylbutane

Stabilized with potassium carbonate

Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 121…122 °C
Density: 1.223
Flash Pt: 22 °C (79 °F)
Storage Temperature: Ambient
CAS Number: 5973-11-5
EINECS: 208-583-5
UN: 1993
ADR: 3 III
Merck Index: 14,601

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR