3-Cyano-1-propylboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile

H27126.03 H27126.06
ALFAH27126.03EA 187 SGD
ALFAH27126.03 ALFAH27126.06
3-Cyano-1-propylboronic acid pinacol ester 96%
3-Cyano-1-propylboronic acid pinacol ester

Purity: 96% CAS No.: 238088-16-9 Molecular Formula: C10H18BNO2 Formula Weight: 195.07 Refractive Index: 1.4370

Formula: C₁₀H₁₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD09953480
CAS Number: 238088-16-9
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR