4-Cyano-3-pyridineboronic acid pinacol ester

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isonicotinonitrile, 4-Cyanopyridine-3-boronic acid, pinacol ester, 4-Cyanopyridine-3-boronic acid pinacol ester

H27216.MD H27216.03
ALFAH27216.MDEA 0 SGD
ALFAH27216.MD ALFAH27216.03
4-Cyano-3-pyridineboronic acid pinacol ester
4-Cyano-3-pyridineboronic acid pinacol ester

CAS No.: 878194-91-3 Molecular Formula: C12H15BN2O2 Formula Weight: 230.07 Melting Point: 84-88° MDL No.: MFCD08458478

Formula: C₁₂H₁₅BN₂O₂
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD08458478
CAS Number: 878194-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR