4'-Chloro-2'-hydroxyacetophenone ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(4-Chloro-2-hydroxyphenyl)ethan-1-one, 4-Chloro-2-hydroxyacetophenone

H27309.18 H27309.09
ALFAH27309.18EA 0 SGD
ALFAH27309.18 ALFAH27309.09
4'-Chloro-2'-hydroxyacetophenone ≥97%
4'-Chloro-2'-hydroxyacetophenone

Purity: 97% CAS No.: 6921-66-0 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 27-30° MDL No.: MFCD00238557

Formula: C₈H₇ClO₂
Melting Pt: 27…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00238557
CAS Number: 6921-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR