3-Bromo-5-chloropyridine 95%

Supplier: Thermo Fisher Scientific
Danger

H27454.06
ALFAH27454.06EA 0 SGD
ALFAH27454.06
3-Bromo-5-chloropyridine 95%
3-Bromo-5-chloropyridine

Purity: 95% CAS No.: 73583-39-8 Molecular Formula: C5H3BrClN Formula Weight: 192.44 Melting Point: 78-81° MDL No.: MFCD04114221

Formula: C₅H₃BrClN
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD04114221
CAS Number: 73583-39-8
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR