4-Iodonicotinonitrile 95%

Supplier: Thermo Fisher Scientific
Danger

H27603.MD H27603.03
ALFAH27603.MDEA 0 SGD
ALFAH27603.MD ALFAH27603.03
4-Iodonicotinonitrile 95%
4-Iodonicotinonitrile

Purity: 95% CAS No.: 490039-72-0 Molecular Formula: C6H3IN2 Formula Weight: 230.01 Storage Temperature: -30°C to -10°C Melting Point: ca 124° dec. UN No.: 3439 MDL No.: MFCD09037453

Formula: C₆H₃IN₂
Melting Pt: ∼ 124 °C (dec.)
MDL Number: MFCD09037453
CAS Number: 490039-72-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR