1-Ethyl-3-methylimidazolium chloride ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

H27651.06 H27651.14
ALFAH27651.06EA 0 SGD
ALFAH27651.06 ALFAH27651.14
1-Ethyl-3-methylimidazolium chloride ≥98%
1-Ethyl-3-methylimidazolium chloride

Purity: 98+% CAS No.: 65039-09-0 Molecular Formula: C6H11ClN2 Formula Weight: 146.62 Flash Point: 186°(367°F) Melting Point: 77-80° MDL No.: MFCD00074843

Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
Melting Pt: 77…80 °C
Density: ∼1.1 g/cm³ (20 °C)
Flash Pt: 186 °C (367 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074843
CAS Number: 65039-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR