4-Bromo-2-fluoropyridine 95%

Supplier: Thermo Fisher Scientific
Danger

H27657.06
ALFAH27657.06EA 0 SGD
ALFAH27657.06
4-Bromo-2-fluoropyridine 95%
4-Bromo-2-fluoropyridine

Purity: 95% CAS No.: 128071-98-7 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Refractive Index: 1.5300

Formula: C₅H₃BrFN
Storage Temperature: Ambient
MDL Number: MFCD04112504
CAS Number: 128071-98-7
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR