4-Amino-5-bromo-2-chloropyridine 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-bromo-2-chloropyridin-4-amine, 5-Bromo-2-chloro-4-pyridylamine, 5-Bromo-2-chloro-4-pyridinamine

H27674.03
ALFAH27674.03EA 0 SGD
ALFAH27674.03
4-Amino-5-bromo-2-chloropyridine 95%
4-Amino-5-bromo-2-chloropyridine

Purity: 95% CAS No.: 857730-21-3 Molecular Formula: C5H4BrClN2 Formula Weight: 207.46 Melting Point: 127-129° MDL No.: MFCD09037459

Formula: C₅H₄BrClN₂
MW: 207.46
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD09037459
CAS Number: 857730-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR