5-Bromo-2-fluoropyrimidine 95%

Supplier: Thermo Fisher Scientific
Warning

H27795.03
ALFAH27795.03EA 0 SGD
ALFAH27795.03
5-Bromo-2-fluoropyrimidine 95%
5-Bromo-2-fluoropyrimidine

Purity: 95% CAS No.: 62802-38-4 Molecular Formula: C4H2BrFN2 Formula Weight: 176.98 Melting Point: 90-93° MDL No.: MFCD07787364

Formula: C₄H₂BrFN₂
MW: 176.98 g/mol
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD07787364
CAS Number: 62802-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR