2',3-Dihydroxyflavone ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Hydroxy-2-(2-hydroxyphenyl)chromen-4-one, 3-Hydroxy-2-(2-hydroxyphenyl)-4-benzopyrone

H27841.03
ALFAH27841.03EA 0 SGD
ALFAH27841.03
2',3-Dihydroxyflavone ≥97%
2',3-Dihydroxyflavone

Purity: 97% CAS No.: 6068-76-4 Molecular Formula: C15H10O4 Formula Weight: 254.24 Melting Point: 201-204° MDL No.: MFCD00017674

Formula: C₁₅H₁₀O₄
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00017674
CAS Number: 6068-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR