1-Bromo-2-hexadecanone ≥97%

Supplier: Thermo Fisher Scientific
Warning

H27872.09
ALFAH27872.09EA 0 SGD
ALFAH27872.09
1-Bromo-2-hexadecanone ≥97%
1-Bromo-2-hexadecanone

Purity: 97% CAS No.: 21436-52-2 Molecular Formula: C16H31BrO Formula Weight: 319.32 Melting Point: 54-59° MDL No.: MFCD07777089

Formula: C₁₆H₃₁BrO
Melting Pt: 54…59 °C
Storage Temperature: Ambient
MDL Number: MFCD07777089
CAS Number: 21436-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR