Ethyl-7-bromoheptanoate 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 7-Bromoheptanoic Acid Ethyl Ester, Ethyl 7-bromoheptanoate

H28001.AA
ALFAH28001.AAEA 0 SGD
ALFAH28001.AA
Ethyl-7-bromoheptanoate 97%
Ethyl-7-bromoheptanoate

Purity: 97% CAS No.: 29823-18-5 Molecular Formula: Br(CH2)6CO2CH2CH3 Formula Weight: 237.14 Flash Point: 110°(230°F) Boiling Point: 112°/5mm MDL No.: MFCD00037103

Formula: C₉H₁₇BrO₂
MW: 237.14 g/mol
Boiling Pt: 112 °C (5 mmHg)
Density: 1.217
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037103
CAS Number: 29823-18-5
EINECS: 249-883-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR