3-Bromo-p-cresol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Bromo-4-methylphenol, 1-Bromo-3-hydroxy-6-methylbenzene

H28689.06 H28689.03
ALFAH28689.06EA 0 SGD
ALFAH28689.06 ALFAH28689.03
3-Bromo-p-cresol ≥98%
3-Bromo-p-cresol

Purity: 98% CAS No.: 60710-39-6 Molecular Formula: C7H7BrO Formula Weight: 187.04 Melting Point: 54-57° MDL No.: MFCD08273793

Formula: C₇H₇BrO
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD08273793
CAS Number: 60710-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR