ω-Bromo-2-nitroacetophenone 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: α-Bromo-2-nitroacetophenone, 2-Bromo-2'-nitroacetophenone, Bromomethyl o-nitrophenyl ketone, 2-Nitrophenacyl bromide

H30275.03 H30275.06
ALFAH30275.03EA 0 SGD
ALFAH30275.03 ALFAH30275.06
ω-Bromo-2-nitroacetophenone 98
ω-Bromo-2-nitroacetophenone

Purity: 98% CAS No.: 6851-99-6 Molecular Formula: C8H6BrNO3 Formula Weight: 244.05 Odour: Pungent Melting Point: 53-56° UN No.: 3261 MDL No.: MFCD00010294

Formula: C₈H₆BrNO₃
MW: 244.04
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00010294
CAS Number: 6851-99-6
UN: 3261
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR