4-Ethylbenzyl alcohol ≥99%

Supplier: Thermo Fisher Scientific

H30459.03 H30459.06
ALFAH30459.03EA 0 SGD
ALFAH30459.03 ALFAH30459.06
4-Ethylbenzyl alcohol ≥99%
4-Ethylbenzyl alcohol

Purity: 99% CAS No.: 768-59-2 Molecular Formula: C9H12O Formula Weight: 136.19 Refractive Index: 1.5275 Boiling Point: 115-117°/9mm MDL No.: MFCD00004666

Formula: C₉H₁₂O
Boiling Pt: 115…117 °C (9 mmHg)
Density: 1,028
Storage Temperature: Ambient
MDL Number: MFCD00004666
CAS Number: 768-59-2
EINECS: 292-198-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR