2-(3-Chlorophenyl)ethylboronic acid pinacol ester ≥97%

Supplier: Thermo Fisher Scientific

H30464.03 H30464.06
ALFAH30464.03EA 0 SGD
ALFAH30464.03 ALFAH30464.06
2-(3-Chlorophenyl)ethylboronic acid pinacol ester ≥97%
2-(3-Chlorophenyl)ethylboronic acid pinacol ester

Purity: 97% Molecular Formula: C14H20BClO2 Formula Weight: 266.58 Refractive Index: 1.4965 MDL No.: MFCD12546188

Formula: C₁₄H₂₀BClO₂
MW: 266.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546188
CAS Number: 1315277-53-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR