2-Amino-5-chlorobenzyl alcohol 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (2-Amino-5-chlorophenyl)methanol

H30531.06 H30531.14
ALFAH30531.06EA 0 SGD
ALFAH30531.06 ALFAH30531.14
2-Amino-5-chlorobenzyl alcohol 98%
2-Amino-5-chlorobenzyl alcohol

Purity: 98% CAS No.: 37585-25-4 Molecular Formula: C7H8ClNO Formula Weight: 157.60 Melting Point: 107-109° MDL No.: MFCD00075163

Formula: C₇H₈ClNO
MW: 157.60 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00075163
CAS Number: 37585-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR