2-Bromo-6-methoxypyridine ≥97%

Supplier: Thermo Fisher Scientific
Warning

H30593.06 H30593.14
ALFAH30593.06EA 0 SGD
ALFAH30593.06 ALFAH30593.14
2-Bromo-6-methoxypyridine ≥97%
2-Bromo-6-methoxypyridine

Purity: 97% CAS No.: 40473-07-2 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Flash Point: 104°(219°F) Refractive Index: 1.5600 Boiling Point: 206° MDL No.: MFCD00088345

Formula: C₆H₆BrNO
Boiling Pt: 206 °C
Density: 1,530
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00088345
CAS Number: 40473-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR