2-Bromo-5-methoxybenzamide ≥98%

Supplier: Thermo Fisher Scientific
Warning

H31250.06 H31250.14
ALFAH31250.06EA 0 SGD
ALFAH31250.06 ALFAH31250.14
2-Bromo-5-methoxybenzamide ≥98%
2-Bromo-5-methoxybenzamide

Purity: 98% CAS No.: 99848-43-8 Molecular Formula: C8H8BrNO2 Formula Weight: 230.66 Melting Point: 159-161° MDL No.: MFCD00208336

Formula: C₈H₈BrNO₂
MW: 230.66 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00208336
CAS Number: 99848-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR