2-Bromo-5-methylbenzotrifluoride ≥98%

Supplier: Thermo Fisher Scientific
Warning

H31523.03
ALFAH31523.03EA 0 SGD
ALFAH31523.03
2-Bromo-5-methylbenzotrifluoride ≥98%
2-Bromo-5-methylbenzotrifluoride

Purity: 98% CAS No.: 261952-20-9 Molecular Formula: C8H6BrF3 Formula Weight: 239.04 MDL No.: MFCD01631586

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631586
CAS Number: 261952-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR