5-Bromo-3-(trifluoromethoxy)salicylaldehyde 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Bromo-2-hydroxy-3-(trifluoromethoxy)benzaldehyde

H31842.03
ALFAH31842.03EA 0 SGD
ALFAH31842.03
5-Bromo-3-(trifluoromethoxy)salicylaldehyde 99%
5-Bromo-3-(trifluoromethoxy)salicylaldehyde

Purity: 99% Molecular Formula: C8H4BrF3O3 Formula Weight: 285.02 Melting Point: 44-48° MDL No.: MFCD03788532

Formula: C₈H₄BrF₃O₃
MW: 285.02 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03788532
CAS Number: 497959-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR