Tris[2-(dimethylamino)ethyl]amine ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: Me6TREN

H31853.06
ALFAH31853.06EA 0 SGD
ALFAH31853.06
Tris[2-(dimethylamino)ethyl]amine ≥99%
Tris[2-(dimethylamino)ethyl]amine

Purity: 99+% CAS No.: 33527-91-2 Molecular Formula: C12H30N4 Formula Weight: 230.40 Refractive Index: 1.4540 Boiling Point: 70-71°/0.5mm UN No.: UN2735 MDL No.: MFCD00015607

Formula: C₁₂H₃₀N₄
MW: 230.40 g/mol
MDL Number: MFCD00015607
CAS Number: 33527-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR