4-Fluoro-3-methoxybenzyl alcohol 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4-Fluoro-3-methoxyphenyl)methanol

H31897.09
ALFAH31897.09EA 0 SGD
ALFAH31897.09
4-Fluoro-3-methoxybenzyl alcohol 95%
4-Fluoro-3-methoxybenzyl alcohol

Purity: 95% CAS No.: 128495-45-4 Molecular Formula: C8H9FO2 Formula Weight: 156.15 Melting Point: 47-51° MDL No.: MFCD00272133

Formula: C₈H₉FO₂
Melting Pt: 47…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00272133
CAS Number: 128495-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR