6-Bromo-2-chlorobenzothiazole 97%

Supplier: Thermo Fisher Scientific
Danger

H31962.03 H31962.06
ALFAH31962.03EA 0 SGD
ALFAH31962.03 ALFAH31962.06
6-Bromo-2-chlorobenzothiazole 97%
6-Bromo-2-chlorobenzothiazole

Purity: 97% CAS No.: 80945-86-4 Molecular Formula: C7H3BrClNS Formula Weight: 248.53 Melting Point: 98-100° MDL No.: MFCD04971822

Formula: C₇H₃BrClNS
MW: 248.53 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD04971822
CAS Number: 80945-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR