2-Bromo-3-fluoropyridine ≥97%

Supplier: Thermo Fisher Scientific
Danger

H31981.03 H31981.06
ALFAH31981.03EA 0 SGD
ALFAH31981.03 ALFAH31981.06
2-Bromo-3-fluoropyridine ≥97%
2-Bromo-3-fluoropyridine

Purity: 97% CAS No.: 40273-45-8 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Refractive Index: 1.5450

Formula: C₅H₃BrFN
MW: 175.99
Storage Temperature: Ambient
MDL Number: MFCD04114128
CAS Number: 40273-45-8
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR