2-Chloro-4-methoxybenzoic acid 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Chloro-p-anisic acid

H32014.03 H32014.06
ALFAH32014.03EA 0 SGD
ALFAH32014.03 ALFAH32014.06
2-Chloro-4-methoxybenzoic acid 98
2-Chloro-4-methoxybenzoic acid

Purity: 98% CAS No.: 21971-21-1 Molecular Formula: C8H7ClO3 Formula Weight: 186.59 MDL No.: MFCD00085943

Formula: C₈H₇ClO₃
MW: 186.59
Storage Temperature: Ambient
MDL Number: MFCD00085943
CAS Number: 21971-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR