2,3-Difluoro-4-hydroxybenzonitrile 95%

Supplier: Thermo Fisher Scientific
Warning

H32231.03
ALFAH32231.03EA 0 SGD
ALFAH32231.03
2,3-Difluoro-4-hydroxybenzonitrile 95%
2,3-Difluoro-4-hydroxybenzonitrile

Purity: 95% CAS No.: 126162-38-7 Molecular Formula: C7H3F2NO Formula Weight: 155.10 Melting Point: 145-149° MDL No.: MFCD00269596

Formula: C₇H₃F₂NO
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00269596
CAS Number: 126162-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR