4-Hydroxynicotinic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H32282.03
ALFAH32282.03EA 0 SGD
ALFAH32282.03
4-Hydroxynicotinic acid ≥98%
4-Hydroxynicotinic acid

Purity: 98% CAS No.: 609-70-1 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Melting Point: 256-259° MDL No.: MFCD00040286

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 256…259 °C
Storage Temperature: Ambient
MDL Number: MFCD00040286
CAS Number: 609-70-1
EINECS: 210-197-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR