2-Bromo-3,5-bis(trifluoromethyl)aniline 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3,5-Bis(trifluoromethyl)-2-bromoaniline

H32722.14
ALFAH32722.14EA 0 SGD
ALFAH32722.14
2-Bromo-3,5-bis(trifluoromethyl)aniline 95%
2-Bromo-3,5-bis(trifluoromethyl)aniline

Purity: 95% CAS No.: 174824-16-9 Molecular Formula: C8H4BrF6N Formula Weight: 308.02 Melting Point: 33-37° UN No.: 2811 MDL No.: MFCD00134419

Formula: C₈H₄BrF₆N
Melting Pt: 33…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00134419
CAS Number: 174824-16-9
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR