Boron trichloride 1 M in dichloromethane stabilised

Supplier: Thermo Fisher Scientific
Danger

H32751.AE H32751.AP
ALFAH32751.AEEA 0 SGD
ALFAH32751.AE ALFAH32751.AP
Boron trichloride 1 M in dichloromethane stabilised
Boron trichloride

CAS No.: 10294-34-5 Molecular Formula: BCl3 Formula Weight: 117.17 Storage Temperature: 2°C to 8°C UN No.: 2927 MDL No.: MFCD00011313

Formula: BCl₃
Density: 1,326
Storage Temperature: Refrigerator
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 2927
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR