Boron trichloride 1 M in hexanes (mixed isomers)

Supplier: Thermo Fisher Scientific
Danger

H32833.AE H32833.AP
ALFAH32833.AEEA 0 SGD
ALFAH32833.AE ALFAH32833.AP
Boron trichloride 1 M in hexanes (mixed isomers)
Boron trichloride

CAS No.: 10294-34-5 Molecular Formula: BCl3 Formula Weight: 117.17 Storage Temperature: 2°C to 8°C UN No.: 3286 MDL No.: MFCD00011313

Formula: BCl₃
Density: 0,738
Storage Temperature: Refrigerator
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 3286
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR