2-(N-Benzyl-N-methylamino)-5-bromopyridine 95%

Supplier: Thermo Fisher Scientific
Warning

H32929.03
ALFAH32929.03EA 0 SGD
ALFAH32929.03
2-(N-Benzyl-N-methylamino)-5-bromopyridine 95%
2-(N-Benzyl-N-methylamino)-5-bromopyridine

Purity: 95% CAS No.: 1187386-36-2 Molecular Formula: C12H11BrN2 Formula Weight: 263.14 MDL No.: MFCD12913988

Formula: C₁₃H₁₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12913988
CAS Number: 1187386-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR