1-Bromo-4-dodecylbenzene ≥98%

Supplier: Thermo Fisher Scientific

H32963.06 H32963.14
ALFAH32963.06EA 0 SGD
ALFAH32963.06 ALFAH32963.14
1-Bromo-4-dodecylbenzene ≥98%
1-Bromo-4-dodecylbenzene

Purity: 98% CAS No.: 126930-72-1 Molecular Formula: C18H29Br Formula Weight: 325.33 Flash Point: 110°(230°F) Refractive Index: 1.5034 Boiling Point: 376° MDL No.: MFCD00191386

Formula: C₁₈H₂₉Br
Boiling Pt: 376 °C
Density: 1,07
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191386
CAS Number: 126930-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR