6-Bromo-4-methylquinoline 96%

Supplier: Thermo Fisher Scientific
Warning

H33298.MD H33298.03
ALFAH33298.MDEA 0 SGD
ALFAH33298.MD ALFAH33298.03
6-Bromo-4-methylquinoline 96%
6-Bromo-4-methylquinoline

Purity: 96% CAS No.: 41037-28-9 Molecular Formula: C10H8BrN Formula Weight: 222.08 Melting Point: 94-95° MDL No.: MFCD09258893

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Melting Pt: 94…95 °C
Storage Temperature: Ambient
MDL Number: MFCD09258893
CAS Number: 41037-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR