1,3-benzothiazol-5-amine 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,3-Benzothiazol-5-amine, 5-Aminobenzo[d]thiazole, 1,3-Benzothiazol-5-amine, 5-Aminobenzothiazole

H33309.03 H33309.06
ALFAH33309.03EA 0 SGD
ALFAH33309.03 ALFAH33309.06
1,3-benzothiazol-5-amine 96%
1,3-benzothiazol-5-amine

Purity: 96% CAS No.: 1123-93-9 Molecular Formula: C7H6N2S Formula Weight: 150.20 Melting Point: 55-59° MDL No.: MFCD04115282

Formula: C₇H₆N₂S
MW: 150.199 g/mol
Melting Pt: 55…59 °C
Storage Temperature: Ambient
MDL Number: MFCD04115282
CAS Number: 1123-93-9
EINECS: 214-381-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR