4-Acetamidobenzyl alcohol ≥97%

Supplier: Thermo Fisher Scientific
Warning

H33454.03 H33454.06
ALFAH33454.03EA 0 SGD
ALFAH33454.03 ALFAH33454.06
4-Acetamidobenzyl alcohol ≥97%
4-Acetamidobenzyl alcohol
Formula: C₉H₁₁NO₂
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00016868
CAS Number: 16375-88-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR