N-Benzyl-6-chloro-4-pyrimidinamine 96%

Supplier: Thermo Fisher Scientific
Warning

ALFAH33723.MEEA 0 SGD
ALFAH33723.ME
N-Benzyl-6-chloro-4-pyrimidinamine 96%
N-Benzyl-6-chloro-4-pyrimidinamine

Purity: 96% CAS No.: 61667-16-1 Molecular Formula: C11H10ClN3 Formula Weight: 219.67 Melting Point: 116-117° MDL No.: MFCD00679879

Formula: C₁₁H₁₀ClN₃
Melting Pt: 116…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00679879
CAS Number: 61667-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR