3-Amino-2-bromo-5-fluoropyridine 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromo-5-fluoropyridin-3-amine, 2-Bromo-5-fluoro-3-pyridylamine

H33811.06
ALFAH33811.06EA 0 SGD
ALFAH33811.06
3-Amino-2-bromo-5-fluoropyridine 97%
3-Amino-2-bromo-5-fluoropyridine

Purity: 97% CAS No.: 884495-03-8 Molecular Formula: C5H4BrFN2 Formula Weight: 191.00 Melting Point: 97-101° MDL No.: MFCD05662413

Formula: C₅H₄BrFN₂
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD05662413
CAS Number: 884495-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR