2-(4-Fluorophenoxy)benzyl bromide 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-(Bromomethyl)-2-(4-fluorophenoxy)benzene

ALFAH33860.MDEA 0 SGD
ALFAH33860.MD
2-(4-Fluorophenoxy)benzyl bromide 95%
2-(4-Fluorophenoxy)benzyl bromide

Purity: 95% Molecular Formula: C13H10BrFO Formula Weight: 281.12 UN No.: UN3261 MDL No.: MFCD15146803

Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD15146803
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR