Benzothiazole-6-carboxamide 96%

Supplier: Thermo Fisher Scientific
Warning

H34216.06
ALFAH34216.06EA 0 SGD
ALFAH34216.06
Benzothiazole-6-carboxamide 96%
Benzothiazole-6-carboxamide

Purity: 96% CAS No.: 101084-51-9 Molecular Formula: C8H6N2OS Formula Weight: 178.21 Melting Point: 245-246° MDL No.: MFCD11643189

Formula: C₈H₆N₂OS
Melting Pt: 245…246 °C
Storage Temperature: Ambient
MDL Number: MFCD11643189
CAS Number: 101084-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR