Methyl-3-bromo-2-fluorobenzoate ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Methyl 3-bromo-2-fluorobenzoate

H36865.06
ALFAH36865.06EA 693 SGD
ALFAH36865.06
Methyl-3-bromo-2-fluorobenzoate ≥98%
Methyl 3-bromo-2-fluorobenzoate
Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD09261255
CAS Number: 206551-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR