2-Chloro-4-iodonicotinic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Chloro-4-iodopyridine-3-carboxylic acid

H50049.03
ALFAH50049.03EA 0 SGD
ALFAH50049.03
2-Chloro-4-iodonicotinic acid ≥98%
2-Chloro-4-iodonicotinic acid

Purity: 98% CAS No.: 544671-78-5 Molecular Formula: C6H3ClINO2 Formula Weight: 283.45 Melting Point: 162-167° MDL No.: MFCD05663520

Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
Melting Pt: 162…167 °C
Storage Temperature: Ambient
MDL Number: MFCD05663520
CAS Number: 544671-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR