5-Bromo-2-formylpyridine ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Bromopyridine-2-carbaldehyde, 5-Bromopicolinaldehyde, 5-Bromopyridine-2-carboxaldehyde

H50154.03 H50154.06
ALFAH50154.03EA 138 SGD
ALFAH50154.03 ALFAH50154.06
5-Bromo-2-formylpyridine ≥99%
5-Bromo-2-formylpyridine

Purity: 99% CAS No.: 31181-90-5 Molecular Formula: C6H4BrNO Formula Weight: 186.01 Melting Point: 94-96° MDL No.: MFCD04112535

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD04112535
CAS Number: 31181-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR