2-Phthalimidoethanesulphonyl chloride 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)ethanesulphonylchloride

H50382.03 H50382.06
ALFAH50382.03EA 198 SGD
ALFAH50382.03 ALFAH50382.06
2-Phthalimidoethanesulphonyl chloride 97%
2-Phthalimidoethanesulphonyl chloride

Purity: 97% CAS No.: 4403-36-5 Molecular Formula: C10H8ClNO4S Formula Weight: 273.70 Melting Point: 156-160° UN No.: UN3261 MDL No.: MFCD01861217

Formula: C₁₀H₈ClNO₄S
MW: 273.7 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD01861217
CAS Number: 4403-36-5
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR