5-Bromo-2-isopropoxybenzonitrile ≥97%

Supplier: Thermo Fisher Scientific
Danger

H50627.03
ALFAH50627.03EA 357 SGD
ALFAH50627.03
5-Bromo-2-isopropoxybenzonitrile ≥97%
5-Bromo-2-isopropoxybenzonitrile

Purity: 97% CAS No.: 515832-52-7 Molecular Formula: C10H10BrNO Formula Weight: 240.10 UN No.: 3439 MDL No.: MFCD02257441

Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02257441
CAS Number: 515832-52-7
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR