Thiophene-2,5-diboronic acid bis(pinacol) ester 97%

Supplier: Thermo Fisher Scientific

Synonyms: 2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene, Thiophene-2,5-diboronic acid Bis(pinacol) ester, 2,2'-(2,5-Thiophenediyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

H51725.03 H51725.06
ALFAH51725.03EA 174 SGD
ALFAH51725.03 ALFAH51725.06
Thiophene-2,5-diboronic acid bis(pinacol) ester 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester

Purity: 97% CAS No.: 175361-81-6 Molecular Formula: C16H26B2O4S Formula Weight: 336.06 Melting Point: 216-220° MDL No.: MFCD09800563

Formula: C₁₆H₂₆B₂O₄S
MW: 336.06 g/mol
Melting Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD09800563
CAS Number: 175361-81-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR