2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene 95%

Supplier: Thermo Fisher Scientific

Synonyms: 2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 9,9-Dioctylfluorene-2,7-diboronic acid bis(pinacol) ester

H51955.03 H51955.06
ALFAH51955.03EA 156 SGD
ALFAH51955.03 ALFAH51955.06
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene 95%
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-di-n-octylfluorene

Purity: 95% CAS No.: 196207-58-6 Molecular Formula: C41H64B2O4 Formula Weight: 642.58 MDL No.: MFCD16294554

Formula: C₄₁H₆₄B₂O₄
MW: 642.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294554
CAS Number: 196207-58-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR