Fmoc-4-iodo-L-phenylalanine 95%

Supplier: Thermo Fisher Scientific

Synonyms: Fmoc-p-iodo-Phe-OH, Fmoc-Phe(4-I)-OH

H51977.03 H51977.06
ALFAH51977.03EA 237 SGD
ALFAH51977.03 ALFAH51977.06
Fmoc-4-iodo-L-phenylalanine 95%
Fmoc-4-iodo-L-phenylalanine

Purity: 95% CAS No.: 82565-68-2 Molecular Formula: C24H20INO4 Formula Weight: 513.33 Melting Point: 213-217° Optical Rotation: -10° (c=1 in DMF) MDL No.: MFCD00672340

Formula: C₂₄H₂₀INO₄
MW: 513.331 g/mol
Melting Pt: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00672340
CAS Number: 82565-68-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR