2-Benzyl-D-proline hydrochloride 95%

Supplier: Thermo Fisher Scientific

H52019.03
ALFAH52019.03EA 988 SGD
ALFAH52019.03
2-Benzyl-D-proline hydrochloride 95%
2-Benzyl-D-proline hydrochloride

Purity: 95% CAS No.: 637020-57-6 Molecular Formula: C12H15NO2.HCl Formula Weight: 241.72 Optical Rotation: +33° (c=1 in 1N HCl) MDL No.: MFCD07367715

Formula: C₁₂H₁₅NO₂·HCl
MW: 241.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367715
CAS Number: 637020-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR